2-Ethyltoluene
2-Ethyltoluene
Catalog # EHERC13356200
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCc1ccccc1C
- Inchi: InChI=1S/C9H12/c1-3-9-7-5-4-6-8(9)2/h4-7H,3H2,1-2H3
- IUPAC: 1-ethyl-2-methylbenzene
- Packing group: III
Specifications
Specifications
- Description:2-Ethyltoluene
- UN No.:3295
- Cat. No.:EHERC13356200
- Formula:C₉H₁₂
- Storage conditions:20 °C
- Hazard class description:GHS02, GHS08
- GHS signal:Danger
- Synonym(s):Benzene;1-ethyl-2-methyl-;Toluene;o-ethyl- (8CI);1-Ethyl-2-methylbenzene;1-Methyl-2-ethylbenzene;2-Ethyltoluene;2-Methyl-1-ethylbenzene;2-Methylethylbenzene;NSC 405731;o-Ethylmethylbenzene;o-Ethyltoluene;o-Methylethylbenzene
- Molecular weight:120,19
- CAS No.:611-14-3
- Shipping conditions:RT



