2,4,6-Trichlorophenol acetate
2,4,6-Trichlorophenol acetate
Catalog # EHERC17775500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(=O)Oc1c(Cl)cc(Cl)cc1Cl
- Inchi: InChI=1S/C8H5Cl3O2/c1-4(12)13-8-6(10)2-5(9)3-7(8)11/h2-3H,1H3
- IUPAC: (2,4,6-trichlorophenyl) acetate
- Packing group: III
Specifications
Specifications
- Description:2,4,6-Trichlorophenol acetate
- UN No.:3077
- Cat. No.:EHERC17775500
- Formula:C₈H₅Cl₃O₂
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Molecular weight:239,48
- CAS No.:23399-90-8
- Shipping conditions:20



