1,5-Dimethylnaphthalene
1,5-Dimethylnaphthalene
Catalog # EHERC20762000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Cc1cccc2c(C)cccc12
- Inchi: InChI=1S/C12H12/c1-9-5-3-8-12-10(2)6-4-7-11(9)12/h3-8H,1-2H3
- IUPAC: 1,5-dimethylnaphthalene
- Packing group: III
Specifications
Specifications
- Description:1,5-Dimethylnaphthalene
- UN No.:3077
- Cat. No.:EHERC20762000
- Formula:C₁₂H₁₂
- Storage temperature:20 °C
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):Naphthalene;1,5-dimethyl- (8CI;9CI;ACI);1,5-Dimethylnaphthalene (ACI);NSC 59388
- Molecular weight:156,22
- CAS No.:571-61-9
- Shipping conditions:RT



