Vancomycin hydrochloride
Vancomycin hydrochloride
Catalog # EHERC17900500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Cl.CNC(CC(C)C)C(=O)NC1C(O)c2ccc(Oc3cc4cc(Oc5ccc(cc5Cl)C(O)C6NC(=O)C(NC(=O)C4NC(=O)C(CC(=O)N)NC1=O)c7ccc(O)c(c7)c8c(O)cc(O)cc8C(NC6=O)C(=O)O)c3O[C@@H]9O[C@H](CO)[C@@H](O)[C@H](O)[C@H]9O[C@H]%10C[C@](C)(N)[C@H](O)[C@H](C)O%10)c(Cl)c2
- Inchi: InChI=1S/C66H75Cl2N9O24.ClH/c1-23(2)12-34(71-5)58(88)76-49-51(83)26-7-10-38(32(67)14-26)97-40-16-28-17-41(55(40)101-65-56(54(86)53(85)42(22-78)99-65)100-44-21-66(4,70)57(87)24(3)96-44)98-39-11-8-27(15-33(39)68)52(84)50-63(93)75-48(64(94)95)31-18-29(79)19-37(81)45(31)30-13-25(6-9-36(30)80)46(60(90)77-50)74-61(91)47(28)73-59(89)35(20-43(69)82)72-62(49)92;/h6-11,13-19,23-24,34-35,42,44,46-54,56-57,65,71,78-81,83-87H,12,20-22,70H2,1-5H3,(H2,69,82)(H,72,92)(H,73,89)(H,74,91)(H,75,93)(H,76,88)(H,77,90)(H,94,95);1H/t24-,34?,35?,42+,44-,46?,47?,48?,49?,50?,51?,52?,53+,54-,56+,57+,65-,66-;/m0./s1
- IUPAC: Vancomycin hydrochloride
Specifications
Specifications
- Description:Vancomycin hydrochloride
- Cat. No.:EHERC17900500
- Formula:C₆₆H₇₅Cl₂N₉O₂₄·ClH
- Storage conditions:–18 °C
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):Vancomycin Hydrochloride; 22H-8,11:18,21-Dietheno-23,36-(iminomethano)-13,16:31,35-dimetheno-1H,16H-[1,6,9]oxadiazacyclohexadecino[4,5-m][10,2,16]benzoxadiazacyclotetracosine, vancomycin deriv.; Laikexin; Lyphocin; Meek; Vancocin hydrochloride; Vancocine hydrochloride; Vancocyn; Vancor
- Molecular weight:1485,71
- CAS No.:1404-93-9
- Shipping conditions:RT



