Urea
Urea
Catalog # EHERC17897350
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: O=C(N)N
- Inchi: InChI=1S/CH4N2O/c2-1(3)4/h(H4,2,3,4)
- IUPAC: urea
Specifications
Specifications
- Description:Urea
- Cat. No.:EHERC17897350
- Formula:CH₄N₂O
- Storage conditions:20 °C
- Synonym(s):Urea; AUS 32; AdBlue; Aquacare; Aquadrate; B-I-K; Basodexan; Benural 70; Carbamide; Carbamimidic acid; Carbonyl diamide; Carmol 40; Cellpaste K 4; Cellton NP; DEF; DEF (diesel exhaust fluid); Diesel exhaust fluid; Duration III; ESN; Elaqua XX; Eucerin 10% Urea Lotion; Hyanit; Isourea; Keratinamin; Keratinamin Kowa; Low Biuret Urea; NSC 34375; Nutraplus; Onychomal; Onyster; Optigen 1200; Optigen II; Pastaron; Pastaron 10; Pastaron 20; Pastaron 20 soft; Pastaron soft; Pseudourea; Rubinol ST 010; SGN 250; UR; Ultra Low Biuret Urea; Urea perhydrate; Ureaphil; Ureophil; Urepeal; Urepeal L; Urepearl; Urevert; Uria; Uroderm; Varioform II
- Molecular weight:60055
- CAS No.:57-13-6
- Shipping conditions:20 °C



