Thimerosal
Thimerosal
Catalog # EHERC17515000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: [Na+].CC[Hg]Sc1ccccc1C(=O)[O-]
- Inchi: InChI=1S/C7H6O2S.C2H5.Hg.Na/c8-7(9)5-3-1-2-4-6(5)10;1-2;;/h1-4,10H,(H,8,9);1H2,2H3;;/q;;2*+1/p-2
- IUPAC: sodium;(2-carboxylatophenyl)sulfanyl-ethylmercury
- Packing group: II
Specifications
Specifications
- Description:Thimerosal
- UN No.:2025
- Cat. No.:EHERC17515000
- Formula:C₉H₉HgO₂S·Na
- Storage conditions:20 °C
- Hazard class description:GHS06, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Thiomersal; Sodium ethyl[2-sulfanylbenzoato(2-)-O,S]mercurate(1-)
- Molecular weight:404,81
- CAS No.:54-64-8
- Shipping conditions:RT



