Tetrasul
Tetrasul
Catalog # EHERC17420000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Clc1ccc(Sc2cc(Cl)c(Cl)cc2Cl)cc1
- Inchi: InChI=1S/C12H6Cl4S/c13-7-1-3-8(4-2-7)17-12-6-10(15)9(14)5-11(12)16/h1-6H
- IUPAC: 1,2,4-trichloro-5-(4-chlorophenyl)sulfanylbenzene
Specifications
Specifications
- Description:Tetrasul
- Cat. No.:EHERC17420000
- Formula:C₁₂H₆Cl₄S
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):Sulfide, p-chlorophenyl 2,4,5-trichlorophenyl (6CI,7CI,8CI); 1,2,4-Trichloro-5-[(4-chlorophenyl)thio]benzene; 2,4,4',5-Tetrachlorodiphenyl sulfide; 4-Chlorophenyl 2,4,5-trichlorophenyl sulfide; Animert; Animert V 101; Tetrasul; V 101; p-Chlorophenyl 2,4,5-trichlorophenyl sulfide
- Molecular weight:324,05
- CAS No.:2227-13-6
- Shipping conditions:RT



