Terbacil
Terbacil
Catalog # EHERC17250000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC1=C(Cl)C(=O)N(C(=O)N1)C(C)(C)C
- Inchi: InChI=1S/C9H13ClN2O2/c1-5-6(10)7(13)12(8(14)11-5)9(2,3)4/h1-4H3,(H,11,14)
- IUPAC: 3-tert-butyl-5-chloro-6-methyl-1H-pyrimidine-2,4-dione
Specifications
Specifications
- Description:Terbacil
- Cat. No.:EHERC17250000
- Formula:C₉H₁₃ClN₂O₂
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Molecular weight:216,66
- CAS No.:5902-51-2
- Shipping conditions:RT



