Propyrisulfuron
Propyrisulfuron
EHERC16537000
- SMILES: CCCc1ccc2nc(Cl)c(n2n1)S(=O)(=O)NC(=O)Nc3nc(OC)cc(OC)n3
- Inchi: InChI=1S/C16H18ClN7O5S/c1-4-5-9-6-7-10-18-13(17)14(24(10)22-9)30(26,27)23-16(25)21-15-19-11(28-2)8-12(20-15)29-3/h6-8H,4-5H2,1-3H3,(H2,19,20,21,23,25)
- IUPAC: 1-(2-chloro-6-propylimidazo[1,2-b]pyridazin-3-yl)sulfonyl-3-(4,6-dimethoxypyrimidin-2-yl)urea
- Packing group: III
- Description:Propyrisulfuron
- UN No.:3077
- Cat. No.:EHERC16537000
- Formula:C₁₆H₁₈ClN₇O₅S
- Storage conditions:–18 °C
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Synonym(s):2-Chloro-N-[[(4,6-dimethoxy-2-pyrimidinyl)amino]carbonyl]-6-propylimidazo[1,2-b]pyridazine-3-sulfonamide; 1-[(2-Chloro-6-propylimidazo[1,2-b]pyridazin-3-yl)sulfonyl]-3-(4,6-dimethoxypyrimidin-2-yl)urea; Propyrisulfuron; TH 547; Zeta-One
- Molecular weight:455,88
- CAS No.:570415-88-2
- Shipping conditions:RT



