Prednisolone
Prednisolone
Catalog # EHERC16286500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@]2(O)C(=O)CO
- Inchi: InChI=1S/C21H28O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h5,7,9,14-16,18,22,24,26H,3-4,6,8,10-11H2,1-2H3/t14-,15-,16-,18+,19-,20-,21-/m0/s1
- IUPAC: (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
Specifications
Specifications
- Description:Prednisolone
- Cat. No.:EHERC16286500
- Formula:C₂₁H₂₈O₅
- Storage conditions:20 °C
- Hazard class description:GHS08
- GHS signal:Danger
- Synonym(s):Hydrocortisone Imp. A (EP); Prednisolone Acetate Imp. B (EP); 11,17,21-Trihydroxy-(11beta)-Pregna-1,4-diene-3,20-dione; 11Beta,17,21-trihydroxy-Pregna-1,4-diene-3,20-dione; Delta1-Cortisol; Delta1-Dehydrocortisol; Delta1-Dehydrohydrocortisone; Delta1-Hydrocortisone; 1,2-Dehydrohydrocortisone; 1,4-Pregnadiene-11beta,17alpha,21-triol-3,20-dione; 1,4-Pregnadiene-3,20-dione-11beta,17alpha,21-triol; 1-Dehydrohydrocortisone; 11beta,17,21-Trihydroxypregna-1,4-diene-3,20-dione; 11beta,17alpha,21-Trihydroxypregna-1,4-diene-3,20-dione; Co-Hydeltra; Codelcortone; Cortalone; Decaprednil; Decortin H; Delcortol; Delta F; Delta-Cortef; Delta-Ef-Cortelan; Delta-stab; Deltacortenol; Deltacortril; Deltacortril Enteric; Deltahydrocortisone; Deltasolone; Deltisilone; Di-Adreson F; Dicortol; Donisolone; Eazolin D; Fernisolone; Flamasone; Hostacortin H; Hydeltra; Hydeltrone; Hydrodeltalone; Hydrodeltisone; Hydroretrocortin; Hydroretrocortine; Klismacort; Metacortandralone; Meti-Derm; Meticortelone; NSC 9120; NSC 9900; Panafcortelone; Paracortol; Precortalon; Precortancyl; Precortilon; Precortisyl; Predne-Dome; Prednelan; Predni-Dome; Prednicen; Predniretard; Prednisolone; Predonin; Predonine; Preflam; Prelone; Prenolone; Scherisolon; Solone; Steran; Sterane; Sterolone; Supercortisol; Ulacort; Ultracorten H; Ultracortene H Prednisolone Acetate Imp. B (EP); Hydrocortisone Impurity A; Methylprednisolone Impurity K; Prednicarbate Impurity A; Prednisolone Acetate Impurity B; Prednisone Impurity B
- Molecular weight:360,44
- CAS No.:50-24-8
- Shipping conditions:RT



