Piperacillin
Piperacillin
Catalog # EHERC16218900
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCN1CCN(C(=O)N[C@@H](C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C(=O)O)c4ccccc4)C(=O)C1=O
- Inchi: InChI=1S/C23H27N5O7S/c1-4-26-10-11-27(19(32)18(26)31)22(35)25-13(12-8-6-5-7-9-12)16(29)24-14-17(30)28-15(21(33)34)23(2,3)36-20(14)28/h5-9,13-15,20H,4,10-11H2,1-3H3,(H,24,29)(H,25,35)(H,33,34)/t13-,14-,15+,20-/m1/s1
- IUPAC: (2S,5R,6R)-6-[[(2R)-2-[(4-ethyl-2,3-dioxopiperazine-1-carbonyl)amino]-2-phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Specifications
Specifications
- Description:Piperacillin
- Cat. No.:EHERC16218900
- Formula:C₂₃H₂₇N₅O₇S
- Storage conditions:4 °C
- Hazard class description:GHS07, GHS08, GHS09
- GHS signal:Warning
- Synonym(s):Piperacillin; (2S,5R,6R)-6-[[(2R)-[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid; [2S-[2alpha,5alpha,6beta(S*)]]-6-[[[[(4-Ethyl-2,3-dioxo-1-piperazinyl)carbonyl]amino]phenylacetyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
- Molecular weight:517,55
- CAS No.:61477-96-1
- Shipping conditions:RT



