Phenol
Phenol
Catalog # EHERC16025000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Oc1ccccc1
- Inchi: InChI=1S/C6H6O/c7-6-4-2-1-3-5-6/h1-5,7H
- IUPAC: phenol
- Packing group: II
Specifications
Specifications
- Description:Phenol
- UN No.:1671
- Cat. No.:EHERC16025000
- Formula:C₆H₆O
- Storage conditions:20 °C
- Hazard class description:GHS05, GHS06, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Phenol; Guaiacol Imp. B (EP); Hexylresorcinol Imp. A (EP); Salicylic Acid Imp. C (EP); 2-Allphenol; Benzenol; Carbolic acid; ENT 1814; Hydroxybenzene; Monohydroxybenzene; Monophenol; NSC 36808; Oxybenzene; Phenic acid; Phenyl alcohol; Phenyl hydrate; Phenyl hydroxide; Phenylic acid; Phenylic alcohol; Guaiacol Impurity B; Hexylresorcinol Impurity A; Methyl Salicylate Impurity B; Salicylic Acid Impurity C
- Molecular weight:94,11
- CAS No.:108-95-2
- Shipping conditions:RT



