Perfluorodecanoic acid
Perfluorodecanoic acid
Catalog # EHERC15986600
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: O=C(O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
- Inchi: InChI=1S/C10HF19O2/c11-2(12,1(30)31)3(13,14)4(15,16)5(17,18)6(19,20)7(21,22)8(23,24)9(25,26)10(27,28)29/h(H,30,31)
- IUPAC: 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanoic acid
- Packing group: III
Specifications
Specifications
- Description:Perfluorodecanoic acid
- UN No.:2811
- Cat. No.:EHERC15986600
- Formula:C₁₀HF₁₉O₂
- Storage conditions:20 °C
- Hazard class description:GHS05, GHS06, GHS07, GHS08
- GHS signal:Danger
- Synonym(s):Decanoic acid, nonadecafluoro- (6CI,7CI,8CI,9CI); 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluorodecanoic acid; Nonadecafluoro-n-decanoic acid; Nonadecafluorodecanoic acid; PFDA; Perfluoro-1-nonanecarboxylic acid; Perfluoro-n-decanoic acid; Perfluorocapric acid; PFC
- Molecular weight:514,08
- CAS No.:335-76-2
- Shipping conditions:RT



