PCB No. 35
PCB No. 35
Catalog # EHERC20003500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Clc1cccc(c1)c2ccc(Cl)c(Cl)c2
- Inchi: InChI=1S/C12H7Cl3/c13-10-3-1-2-8(6-10)9-4-5-11(14)12(15)7-9/h1-7H
- IUPAC: 1,2-dichloro-4-(3-chlorophenyl)benzene
- Packing group: II
Specifications
Specifications
- Description:PCB No. 35
- UN No.:3432
- Cat. No.:EHERC20003500
- Formula:C₁₂H₇Cl₃
- Storage conditions:20 °C
- Hazard class description:GHS08, GHS09
- GHS signal:Warning
- Synonym(s):3,3',4-Trichloro-1,1'-biphenyl; 3,3',4-Trichlorobiphenyl; 3,4,3'-Trichlorobiphenyl; CB 35; PCB 35; 3,3',4-TriCB
- Molecular weight:257,54
- CAS No.:37680-69-6
- Shipping conditions:RT



