Order Entry
Export
Orders LinkContactUsLinkComponent
 

N-Nitrosopiperidine

N-Nitrosopiperidine
  EHERC15606500
N-Nitrosopiperidine
  EHERC15606500
 :  
N-Nitrosopiperidine
N-Nitrosopiperidine

N-Nitrosopiperidine

 :  
 
112,00 €
   
1 * 25 mg
 
  • SMILES: O=NN1CCCCC1
  • Inchi: InChI=1S/C5H10N2O/c8-6-7-4-2-1-3-5-7/h1-5H2
  • IUPAC: 1-nitrosopiperidine
  • Packing group: III

 

 

 

  • Description:
    N-Nitrosopiperidine
  • UN No.:
    2810
  • Cat. No.:
    EHERC15606500
  • Formula:
    C₅H₁₀N₂O
  • Storage conditions:
    4 °C
  • Hazard class description:
    GHS06, GHS08
  • GHS signal:
    Danger
  • Synonym(s):
    Piperidine, 1-nitroso-; 1-Nitrosopiperidine; N-Nitrosopiperidine; NPI; NPIP; NSC 138
  • Molecular weight:
    114,15
  • CAS No.:
    100-75-4
  • Shipping conditions:
    RT