Nevirapine
Nevirapine
Catalog # EHERC15505500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: Cc1ccnc2N(C3CC3)c4ncccc4C(=O)Nc12
- Inchi: InChI=1S/C15H14N4O/c1-9-6-8-17-14-12(9)18-15(20)11-3-2-7-16-13(11)19(14)10-4-5-10/h2-3,6-8,10H,4-5H2,1H3,(H,18,20)
- IUPAC: 11-cyclopropyl-4-methyl-5H-dipyrido[2,3-b:3',2'-e][1,4]diazepin-6-one
Specifications
Specifications
- Description:Nevirapine
- Cat. No.:EHERC15505500
- Formula:C₁₅H₁₄N₄O
- Storage conditions:4 °C
- Hazard class description:GHS07, GHS08
- GHS signal:Warning
- Synonym(s):Nevirapine Anhydrous; 6H-Dipyrido[3,2-b:2',3'-e][1,4]diazepin-6-one, 11-cyclopropyl-5,11-dihydro-4-methyl- (9CI); BI-RG 587; NSC 641530; Nevarapine; Nevirapine; Viramune
- Molecular weight:266,3
- CAS No.:129618-40-2
- Shipping conditions:20 °C



