Neomycin sulfate
Neomycin sulfate
Catalog # EHERC15500900
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: OC[C@@H]1[C@@H](O[C@@H]2[C@H](N)[C@@H](O)[C@H](O)[C@H](CN)O2)[C@@H](O)[C@H](O[C@H]3[C@H](O[C@@H]4[C@H](N)[C@@H](O)[C@H](O)[C@@H](CN)O4)[C@@H](N)C[C@@H](N)[C@@H]3O)O1.O=S(O)(O)=O.O=S(O)(O)=O.O=S(O)(O)=O.O
Specifications
Specifications
- Description:Neomycin sulfate
- Cat. No.:EHERC15500900
- Formula:C₂₃H₄₆N₆O₁₃
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08
- GHS signal:Danger
- Synonym(s):Neomycin, sulfate (salt); Biosol; Bykomycin; Endomixin; Fradiomycin sulfate; Lidamycin Creme; Myacine; Mycerin sulfate; Mycifradin; Mycifradin N; Myciguent; Neo-Mantle Creme; Neobrettin; Neofracin; Neolate; Neomix; Neomycin sulfate; Neomycin sulphate; Neosulf; New France F; Nivemycin; Otobiotic; Quintess-N; Tuttomycin
- Molecular weight:926,88
- CAS No.:1405-10-3
- Shipping conditions:RT



