HCH (technical)
HCH (technical)
Catalog # EHERC14070000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: ClC1C(Cl)C(Cl)C(Cl)C(Cl)C1Cl
- Inchi: InChI=1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H
- IUPAC: 1,2,3,4,5,6-hexachlorocyclohexane
- Packing group: III
Specifications
Specifications
- Description:HCH (technical)
- UN No.:2761
- Cat. No.:EHERC14070000
- Formula:C₆H₆Cl₆
- Storage conditions:20 °C
- Hazard class description:GHS06, GHS07, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):1,2,3,4,5,6-Hexachlorocyclohexane; BHC; HCH; NSC 11807; NSC 7909; NSC 8093
- Molecular weight:290,83
- CAS No.:608-73-1
- Shipping conditions:RT



