Fluenetil
Fluenetil
Catalog # EHERC13710000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: FCCOC(=O)Cc1ccc(cc1)c2ccccc2
- Inchi: InChI=1S/C16H15FO2/c17-10-11-19-16(18)12-13-6-8-15(9-7-13)14-4-2-1-3-5-14/h1-9H,10-12H2
- IUPAC: 2-fluoroethyl 2-(4-phenylphenyl)acetate
- Packing group: I
Specifications
Specifications
- Description:Fluenetil
- UN No.:2588
- Cat. No.:EHERC13710000
- Formula:C₁₆H₁₅FO₂
- Storage conditions:20 °C
- Hazard class description:GHS06
- GHS signal:Danger
- Synonym(s):4-Biphenylacetic acid, 2-fluoroethyl ester (8CI); Acetic acid, 4-biphenylyl-, 2-fluoroethyl ester (7CI); Ethanol, 2-fluoro-, 4-biphenylacetate (8CI); Fluenethyl; Fluenetil; Lambrol; M 2060; Mytrol; TH 367-1; β-Fluoroethyl 4-biphenylacetate
- Molecular weight:258,29
- CAS No.:4301-50-2
- Shipping conditions:RT



