Flucythrinate
Flucythrinate
Catalog # EHERC13700000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(C)C(C(=O)OC(C#N)c1cccc(Oc2ccccc2)c1)c3ccc(OC(F)F)cc3
- Inchi: InChI=1S/C26H23F2NO4/c1-17(2)24(18-11-13-21(14-12-18)32-26(27)28)25(30)33-23(16-29)19-7-6-10-22(15-19)31-20-8-4-3-5-9-20/h3-15,17,23-24,26H,1-2H3
- IUPAC: [cyano-(3-phenoxyphenyl)methyl] 2-[4-(difluoromethoxy)phenyl]-3-methylbutanoate
- Packing group: III
Specifications
Specifications
- Description:Flucythrinate
- UN No.:3352
- Cat. No.:EHERC13700000
- Formula:C₂₆H₂₃F₂NO₄
- Storage conditions:4 °C
- Hazard class description:GHS06, GHS07, GHS09
- GHS signal:Danger
- Molecular weight:451,46
- CAS No.:70124-77-5
- Shipping conditions:RT



