Fluconazole
Fluconazole
Catalog # EHERC13697500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: OC(Cn1cncn1)(Cn2cncn2)c3ccc(F)cc3F
- Inchi: InChI=1S/C13H12F2N6O/c14-10-1-2-11(12(15)3-10)13(22,4-20-8-16-6-18-20)5-21-9-17-7-19-21/h1-3,6-9,22H,4-5H2
- IUPAC: 2-(2,4-difluorophenyl)-1,3-bis(1,2,4-triazol-1-yl)propan-2-ol
Specifications
Specifications
- Description:Fluconazole
- Cat. No.:EHERC13697500
- Formula:C₁₃H₁₂F₂N₆O
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Biozolene; Diflucan; Elazor; Fluconazole; Flucostat; Flumycon; Flunazol; Flusol; Fluzon; Triflucan; UK 49858; Zoltec; 1H-1,2,4-Triazole-1-ethanol, alpha-(2,4-difluorophenyl)-alpha-(1H-1,2,4-triazol-1-ylmethyl)-
- Molecular weight:306,27
- CAS No.:86386-73-4
- Shipping conditions:RT



