Order Entry
Export
Orders LinkContactUsLinkComponent
 

Fenpyrazamine

Fenpyrazamine
  EHERC13544000
Fenpyrazamine
  EHERC13544000
 :  
Fenpyrazamine
Fenpyrazamine

Fenpyrazamine

 :  
 
425,00 €
   
1 * 100 mg
 
  • SMILES: CC(C)N1N(C(=O)SCC=C)C(=C(C1=O)c2ccccc2C)N
  • Inchi: InChI=1S/C17H21N3O2S/c1-5-10-23-17(22)20-15(18)14(16(21)19(20)11(2)3)13-9-7-6-8-12(13)4/h5-9,11H,1,10,18H2,2-4H3
  • IUPAC: S-prop-2-enyl 5-amino-4-(2-methylphenyl)-3-oxo-2-propan-2-ylpyrazole-1-carbothioate
  • Packing group: III

 

 

 

  • Description:
    Fenpyrazamine
  • UN No.:
    3077
  • Cat. No.:
    EHERC13544000
  • Formula:
    C₁₇H₂₁N₃O₂S
  • Storage conditions:
    20 °C
  • Hazard class description:
    GHS07, GHS09
  • GHS signal:
    Warning
  • Synonym(s):
    1H-Pyrazole-1-carbothioic acid, 5-amino-2,3-dihydro-2-(1-methylethyl)-4-(2-methylphenyl)-3-oxo-, S-2-propen-1-yl ester ; 1H-Pyrazole-1-carbothioic acid, 5-amino-2,3-dihydro-2-(1-methylethyl)-4-(2-methylphenyl)-3-oxo-, S-2-propenyl ester (9CI); 1-[(2-Propenylthio)carbonyl]-2-(1-methylethyl)-4-(2-methylphenyl)-5-amino-1H-pyrazol-3-one; Fenpyrazamine; S 2188; S 2188 (herbicide)
  • Molecular weight:
    331,43
  • CAS No.:
    473798-59-3
  • Shipping conditions:
    RT