Emamectin benzoate
Emamectin benzoate
- SMILES: CC[C@H](C)[C@H]1O[C@@]2(C[C@@H]3C[C@@H](C\C=C(/C)\[C@@H](O[C@H]4C[C@H](OC)[C@@H](O[C@H]5C[C@H](OC)[C@H](NC)[C@H](C)O5)[C@H](C)O4)[C@@H](C)\C=C\C=C\6/CO[C@@H]7[C@H](O)C(=C[C@@H](C(=O)O3)[C@]67O)C)O2)C=C[C@@H]1C.CN[C@@H]8[C@H](C)O[C@H](C[C@@H]8OC)O[C@H]9[C@H](C)O[C@H](C[C@@H]9OC)O[C@H]%10[C@@H](C)\C=C\C=C\%11/CO[C@@H]%12[C@H](O)C(=C[C@@H](C(=O)O[C@H]%13C[C@@H](C\C=C\%10/C)O[C@@]%14(C%13)O[C@H](C(C)C)[C@@H](C)C=C%14)[C@]%11%12O)C.OC(=O)c%15ccccc%15.OC(=O)c%16ccccc%16
- Inchi: InChI=1S/C49H75NO13.C48H73NO13.2C7H6O2/c1-12-26(2)44-29(5)18-19-48(63-44)24-35-21-34(62-48)17-16-28(4)43(27(3)14-13-15-33-25-56-46-42(51)30(6)20-36(47(52)59-35)49(33,46)53)60-40-23-38(55-11)45(32(8)58-40)61-39-22-37(54-10)41(50-9)31(7)57-39;1-25(2)42-28(5)17-18-47(62-42)23-34-20-33(61-47)16-15-27(4)43(26(3)13-12-14-32-24-55-45-41(50)29(6)19-35(46(51)58-34)48(32,45)52)59-39-22-37(54-11)44(31(8)57-39)60-38-21-36(53-10)40(49-9)30(7)56-38;2*8-7(9)6-4-2-1-3-5-6/h13-16,18-20,26-27,29,31-32,34-46,50-51,53H,12,17,21-25H2,1-11H3;12-15,17-19,25-26,28,30-31,33-45,49-50,52H,16,20-24H2,1-11H3;2*1-5H,(H,8,9)/b14-13+,28-16+,33-15+;13-12+,27-15+,32-14+;;/t26-,27-,29-,31-,32-,34+,35-,36-,37-,38-,39-,40-,41+,42+,43-,44+,45-,46+,48+,49+;26-,28-,30-,31-,33+,34-,35-,36-,37-,38-,39-,40+,41+,42+,43-,44-,45+,47+,48+;;/m00../s1
- Packing group: III
- Description:Emamectin benzoate
- UN No.:2811
- Cat. No.:EHERC13117000
- Formula:C₄₉H₇₅NO₁₃·C₄₈H₇₃NO₁₃·2C₇H₆O₂
- Storage conditions:–18 °C
- Hazard class description:GHS05, GHS06, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Avermectin B1, 4''-deoxy-4''-(methylamino)-, (4''R)-, benzoate (1:1); Avermectin B1, 4''-deoxy-4''-(methylamino)-, (4''R)-, benzoate (salt) (9CI); (4''R)-4''-Deoxy-4''-(methylamino)avermectin B1 benzoate; Affirm; Denim; Denim (pesticide); Emamectin benzoate; Emamectin benzoate (salt); MK 244; Methylamino abamectin benzoate; Proclaim; Proclaim 05SG; Proclaim 5SG; Revive; Revive (insecticide); Sch 58854; Shot Wan Liquid Formulation; Shot-Wan; Slice; Tree-age
- Molecular weight:2002,45
- CAS No.:155569-91-8
- Shipping conditions:RT



