Dicumarol
Dicumarol
Catalog # EHERC12581000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: OC1=C(CC2=C(O)c3ccccc3OC2=O)C(=O)Oc4ccccc14
- Inchi: InChI=1S/C19H12O6/c20-16-10-5-1-3-7-14(10)24-18(22)12(16)9-13-17(21)11-6-2-4-8-15(11)25-19(13)23/h1-8,20-21H,9H2
- IUPAC: 4-hydroxy-3-[(4-hydroxy-2-oxochromen-3-yl)methyl]chromen-2-one
- Packing group: III
Specifications
Specifications
- Description:Dicumarol
- UN No.:2811
- Cat. No.:EHERC12581000
- Formula:C₁₉H₁₂O₆
- Storage conditions:4 °C
- Hazard class description:GHS06, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Coumarin, 3,3'-methylenebis[4-hydroxy- ; 3,3'-Methylenebis[4-hydroxy-1,2-benzopyrone]; 3,3'-Methylenebis[4-hydroxycoumarin]; Acadyl; Acavyl; Antitrombosin; Baracoumin; Bis(4-hydroxycoumarin-3-yl)methane; Bis-3,3'-(4-hydroxycoumarinyl)methane; Bishydroxycoumarin; Cuma; Cumid; Di-4-hydroxy-3,3'-methylenedicoumarin; Dicoumal; Dicoumarin; Dicoumarol; Dicuman; Dicumarine; Dicumarol; Dicumol; Dufalone; Kumoran; Melitoxin; NC 034; NSC 17860; NSC 221570; NSC 41834; Temparin; Trombosan
- Molecular weight:336,3
- CAS No.:66-76-2
- Shipping conditions:RT



