Cyprosulfamide
Cyprosulfamide
Catalog # EHERC11915000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: COc1ccccc1C(=O)NS(=O)(=O)c2ccc(cc2)C(=O)NC3CC3
- Inchi: InChI=1S/C18H18N2O5S/c1-25-16-5-3-2-4-15(16)18(22)20-26(23,24)14-10-6-12(7-11-14)17(21)19-13-8-9-13/h2-7,10-11,13H,8-9H2,1H3,(H,19,21)(H,20,22)
- IUPAC: N-[4-(cyclopropylcarbamoyl)phenyl]sulfonyl-2-methoxybenzamide
Specifications
Specifications
- Description:Cyprosulfamide
- Cat. No.:EHERC11915000
- Formula:C₁₈H₁₈N₂O₅S
- Storage conditions:20 °C
- Molecular weight:374,41
- CAS No.:221667-31-8
- Shipping conditions:RT



