Carprofen
Carprofen
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(C(=O)O)c1ccc2c(c1)[nH]c3ccc(Cl)cc23
- Inchi: InChI=1S/C15H12ClNO2/c1-8(15(18)19)9-2-4-11-12-7-10(16)3-5-13(12)17-14(11)6-9/h2-8,17H,1H3,(H,18,19)
- IUPAC: 2-(6-chloro-9H-carbazol-2-yl)propanoic acid
- Packing group: III
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:Carprofen
- UN No.:2811
- Cat. No.:EHERC11045850
- Formula:C₁₅H₁₂ClNO₂
- Storage conditions:20 °C
- Hazard class description:GHS06
- GHS signal:Danger
- Synonym(s):Carprofen; (2RS)-2-(6-Chloro-9H-carbazol-2-yl)propanoic acid
- Molecular weight:273,71
- CAS No.:53716-49-7
- Shipping conditions:RT



