Carbendazim
Carbendazim
Catalog # EHERC10990000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: COC(=O)Nc1nc2ccccc2[nH]1
- Inchi: InChI=1S/C9H9N3O2/c1-14-9(13)12-8-10-6-4-2-3-5-7(6)11-8/h2-5H,1H3,(H2,10,11,12,13)
- IUPAC: methyl N-(1H-benzimidazol-2-yl)carbamate
- Packing group: III
Specifications
Specifications
- Description:Carbendazim
- UN No.:3077
- Cat. No.:EHERC10990000
- Formula:C₉H₉N₃O₂
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Albendazole Imp. E (EP); Fenbendazole Imp. A (EP); Fenbendazole USP Related Compound A; Fenbendazole USP RC A; Carbendazim; Methyl (1H-Benzimidazol-2-yl)carbamate; 1H-Benzimidazol-2-ylcarbamic acid methyl ester; 2-Benzimidazolecarbamic acid methyl ester; 2-(Carbomethoxyamino)benzimidazole; 2-(Methoxycarbamoyl)benzimidazole; 2-(Methoxycarboxamido)benzimidazole; 2-[(Methoxycarbonyl)amino]benzimidazole; A 118; A 118 (pesticide); Agrizim; Antibac MF; BAS 3460; BAS 3460F; BAS 67054F; BCM; BCM (fungicide); BMK; BMK (fungicide); Battal; Bavistan; Bavistin; Bavistin 25SD; Bavistin 3460; Bavistin 50SD; Bavistin FL; Bavistine; Bengard; Bercema-Bitosen; Bitosen; CTR 6669; Carben VL; Carbendazime; Carbendazol; Carbendazole; Delsene; Delsene 10; Derosal; Derosal 60PM; EK 578; Falicarben; Funaben; Funaben 3; Funaben 50; Fungoxan; G 665; Garbenda; HOE 17411; IPO 1250; IPO Y; Jkstein; Karben; Karben Stefes Flo; Karben flo Stefes; Kolfugo; Kolfugo 25FW; Kolfugo Extra; Lignasan; MBC; Mecarzole; Medamine; Methoxycarbonylaminobenzimidazole; Methyl 1H-benzimidazol-2-ylcarbamate; Methyl 1H-benzimidazole-2-carbamate; Methyl 2-benzimidazolecarbamate; Methyl 2-benzimidazolylcarbamate; Methyl benzimidazolylcarbamate; Myco; NSC 109874; Olgin; Olgin (fungicide); Preparation G 665; Preventol BCM; Protek; Sarfun; Spin; Spin (pesticide); Stein; Subeej; Supercarb; Thicoper; Triticol; U 32104; Zhiweiling; SKF-26058; Ph Eur Albendazole Impurity E; Fenbendazole Related Compound A; Albendazole Impurity E; Fenbendazole Impurity A
- Molecular weight:191,19
- CAS No.:10605-21-7
- Shipping conditions:RT



