Bromoethane
Bromoethane
Catalog # EHERC10728000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCBr
- Inchi: InChI=1S/C2H5Br/c1-2-3/h2H2,1H3
- IUPAC: bromoethane
- Packing group: II
Specifications
Specifications
- Description:Bromoethane
- UN No.:1891
- Cat. No.:EHERC10728000
- Formula:C₂H₅Br
- Storage conditions:4 °C
- Hazard class description:GHS02, GHS07, GHS08
- GHS signal:Danger
- Synonym(s):Bromoethane; 2-Bromoethane; Bromic ether; Ethyl bromide; F 160B1; Hydrobromic ether; Monobromoethane; NSC 8824
- Molecular weight:108,97
- CAS No.:74-96-4
- Shipping conditions:RT



