Bisphenol AP
Bisphenol AP
Catalog # EHERC10655640
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(c1ccccc1)(c2ccc(O)cc2)c3ccc(O)cc3
- Inchi: InChI=1S/C20H18O2/c1-20(15-5-3-2-4-6-15,16-7-11-18(21)12-8-16)17-9-13-19(22)14-10-17/h2-14,21-22H,1H3
- IUPAC: 4-[1-(4-hydroxyphenyl)-1-phenylethyl]phenol
- Packing group: III
Specifications
Specifications
- Description:Bisphenol AP
- UN No.:3077
- Cat. No.:EHERC10655640
- Formula:C₂₀H₁₈O₂
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Phenol, 4,4'-(α-methylbenzylidene)di- (6CI,7CI,8CI); 4,4'-(1-Phenylethylidene)bis[phenol]; 1,1-Bis(4-hydroxyphenyl)-1-phenylethane; 1-Phenyl-1,1-bis(4-hydroxyphenyl)ethane; 4,4'-(1-Phenylethylidene)diphenol; 4,4'-(α-Methylbenzylidene)diphenol; Bis(4-hydroxyphenyl)methylphenylmethane; Bisp-Ap; Bisphenol ACP; Bisphenol AP; NK-AP; α,α-Bis(4-hydroxyphenyl)ethylbenzene
- Molecular weight:290,36
- CAS No.:1571-75-1
- Shipping conditions:RT



