Benzene
Benzene
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: c1ccccc1
- Inchi: InChI=1S/C6H6/c1-2-4-6-5-3-1/h1-6H
- IUPAC: benzene
- Packing group: II
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:Benzene
- UN No.:1114
- Cat. No.:EHERC10535000
- Formula:C₆H₆
- Storage conditions:20 °C
- Hazard class description:GHS02, GHS07, GHS08
- GHS signal:Danger
- Synonym(s):Benzene; 1,3,5-Cyclohexatriene; Benzol; Benzole; Coal naphtha; Cyclohexatriene; NSC 67315; Phene; Phenyl hydride; Pyrobenzol; Pyrobenzole; [6]Annulene
- Molecular weight:78,11
- CAS No.:71-43-2
- Shipping conditions:RT



