Acid Red 52
Acid Red 52
Catalog # EHERC10028820
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: [Na+].CCN(CC)c1ccc2c(c3ccc(cc3S(=O)(=O)[O-])S(=O)(=O)[O-])c4ccc(cc4[o+]c2c1)N(CC)CC
- Inchi: InChI=1S/C27H30N2O7S2.Na/c1-5-28(6-2)18-9-12-21-24(15-18)36-25-16-19(29(7-3)8-4)10-13-22(25)27(21)23-14-11-20(37(30,31)32)17-26(23)38(33,34)35;/h9-17H,5-8H2,1-4H3,(H-,30,31,32,33,34,35);/q;+1/p-1
- IUPAC: sodium;4-[3,6-bis(diethylamino)xanthen-10-ium-9-yl]benzene-1,3-disulfonate
Specifications
Specifications
- Description:Acid Red 52
- Cat. No.:EHERC10028820
- Formula:C₂₇H₂₉N₂O₇S₂·Na
- Storage conditions:20 °C
- Synonym(s):3,6-Bis(diethylamino)-9-(2,4-disulfophenyl)xanthylium inner salt sodium salt (1:1); [6-(Diethylamino)-9-(2,4-disulfophenyl)-3H-xanthen-3-ylidene]diethylammonium hydroxide inner salt sodium salt; C.I. Acid Red 52; 3,6-bis(Diethylamino)-9-(2,4-disulfophenyl)xanthylium inner salt sodium salt; Acid Leather Red KB; Acid Red 52; Acid Red B-SF; Acid Red XB; Acid Rhodamine; Acid Rhodamine B; Acid Rose B; Aizen Food Red No. 106; Amacid Rhodamine B; Amido Rhodamine B; Best Acid Rhodamine FB; Brilliant Acid Rhodamine B; Brilliant Superlan Rhodamine 2B; Brilliant Superlan Rhodamine B; C.I. 45100; Colocid Rhodamine B; Colocid Rhodamine BH; Colocid Rhodamine BN; Conacid Red EB; Covasol Red W 4002; Daiwa IJ Red 207H; Daiwa Red 106WB; Dinacid Rhodamine B; Duasyn Acid Rhodamine B 01; Duramine Rhodamine B; Dyacid Red 4B; Dycosacid Rhodamine B; Erio Acid Red XB; Fenazo Pink XXB; Food Color Red No. 106; Food Red 106; Food Red No. 106; Hispacid Brilliant Pink B; Indacid Rhodamine B; Japan Food Red No. 106; Japan Red 106; Japan Red No. 106; KR 620; Kayacyl Rhodamine FB; Kayaku Acid Rhodamine BH; Kayaku Acid Rhodamine FB; Kemacid Rhodamine B; Kiton Red 620; Kiton Red S; Kiton Rhodamine B; Kyton Red; Libacid Rhodamine LB; Lissamine Rhodamine B; Lissamine Rhodamine B 200; Neolan Red E-XB; Nylosan Rhodamine B; Orient Water Red 27; Pacid Rhodamine B; Phloxine rhodamine; Pontacyl Brilliant Pink; Pro-Jet Red OAM; Rakuto Acid Rhodamine; Ravi Acid Rhodamine B; Red 106; Red no. 106; Rhodamine 200S; Rhodamine Acid; Rhodamine B; San-Ei Acid Red; Sandolan Rhodamine E-B; Sandolan Rhodamine E-B 400; Sanolin Rhodamine B; Simacid Red 23011; Solar Rhodamine B; Sulfacid Brilliant Pink 3B; Sulfacid Brilliant Pink 3B 1150; Sulforhodamin B; Sulforhodamine B; Triacid Rhodamine B; Vibra Color Red ARE 52; Vicoacid Red 52Y; Xylene Red B
- Molecular weight:580,65
- CAS No.:3520-42-1
- Shipping conditions:RT



