Acetone-2,4-dinitrophenylhydrazone
Acetone-2,4-dinitrophenylhydrazone
Catalog # EHERC10019500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CC(=NNc1ccc(cc1[N+](=O)[O-])[N+](=O)[O-])C
- Inchi: InChI=1S/C9H10N4O4/c1-6(2)10-11-8-4-3-7(12(14)15)5-9(8)13(16)17/h3-5,11H,1-2H3
- IUPAC: 2,4-dinitro-N-(propan-2-ylideneamino)aniline
- Packing group: III
Specifications
Specifications
- Description:Acetone-2,4-dinitrophenylhydrazone
- UN No.:1325
- Cat. No.:EHERC10019500
- Formula:C₉H₁₀N₄O₄
- Storage conditions:20 °C
- Hazard class description:GHS02, GHS07
- GHS signal:Warning
- Synonym(s):Acetone-2,4-dinitrophenylhydrazone
- Molecular weight:238,2
- CAS No.:1567-89-1
- Shipping conditions:RT



