2-Methoxy-3,5-dimethylpyrazine
2-Methoxy-3,5-dimethylpyrazine
Catalog # EHERC15069000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: COc1ncc(C)nc1C
- Inchi: InChI=1S/C7H10N2O/c1-5-4-8-7(10-3)6(2)9-5/h4H,1-3H3
- IUPAC: 2-methoxy-3,5-dimethylpyrazine
Specifications
Specifications
- Description:2-Methoxy-3,5-dimethylpyrazine
- Cat. No.:EHERC15069000
- Formula:C₇H₁₀N₂O
- Storage conditions:20 °C
- Hazard class description:GHS05, GHS07
- GHS signal:Warning
- Molecular weight:138,17
- CAS No.:92508-08-2
- Shipping conditions:RT



