2-Chloro-5-methylphenol
2-Chloro-5-methylphenol
EHERC11439300
- SMILES: Cc1ccc(Cl)c(O)c1
- Inchi: InChI=1S/C7H7ClO/c1-5-2-3-6(8)7(9)4-5/h2-4,9H,1H3
- IUPAC: 2-chloro-5-methylphenol
- Packing group: II
- Description:2-Chloro-5-methylphenol
- UN No.:2020
- Synonym(s):m-Cresol, 6-chloro- (7CI,8CI); 2-Chloro-5-methylphenol; 1-Chloro-2-hydroxy-4-methylbenzene; 3-Methyl-6-chlorophenol; 4-Chloro-3-hydroxytoluene; 5-Methyl-2-chlorophenol; 6-Chloro-3-methylphenol; 6-Chloro-m-cresol
- Molecular weight:142,58
- CAS No.:615-74-7
- Formula:C₇H₇ClO
- Shipping conditions:RT
- Hazard class description:GHS07, GHS09
- GHS signal:Warning
- Cat. No.:EHERC11439300
- Storage conditions:20 °C



