1-(3,4-Dichlorophenyl)urea
1-(3,4-Dichlorophenyl)urea
Catalog # EHERC12472000
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: NC(=O)Nc1ccc(Cl)c(Cl)c1
- Inchi: InChI=1S/C7H6Cl2N2O/c8-5-2-1-4(3-6(5)9)11-7(10)12/h1-3H,(H3,10,11,12)
- IUPAC: (3,4-dichlorophenyl)urea
Specifications
Specifications
- Description:1-(3,4-Dichlorophenyl)urea
- Cat. No.:EHERC12472000
- Formula:C₇H₆Cl₂N₂O
- Storage conditions:20 °C
- Hazard class description:GHS07
- GHS signal:Warning
- Synonym(s):Urea, N-(3,4-dichlorophenyl)-; Urea, (3,4-dichlorophenyl)- (6CI,7CI,8CI,9CI); N-(3,4-Dichlorophenyl)urea; 1-(3,4-Dichlorophenyl)urea; 3,4-DCPU; 3,4-Dichlorophenylurea; DCPU; Didemethyldiuron; Monouron
- Molecular weight:205,04
- CAS No.:2327-02-8
- Shipping conditions:RT



