Orders LinkContactUsLinkComponent
OptiMATE Fluidics Kit
OptiMATE Fluidics Kit
OptiMATE fluidics kit
Catalog # BECMD24152
New Product
Supplier:   Beckman Coulter
OptiMATE Fluidics Kit
Catalog # BECMD24152
Supplier:  Beckman Coulter
New Product
OptiMATE Fluidics Kit
OptiMATE Fluidics Kit
OptiMATE fluidics kit
Price
1 034,00 €
Log in for availability
1 * 1 items
Product Details & Documents
The OptiMATE Fluidics Kit contains a complete set of consumables needed for gradient dispensing.
Delivery information: The kit includes: 1×upper fluidics, 2×lower fluidics general, 2×lower fluidics conical/BEC gradient replacement fluidics, 1×calibration tube, 1×flush kit, 1×probe guide, 1×seal former and 1×dispense probe.
Specifications
  • Description:
    OptiMATE fluidics kit