Orders LinkContactUsLinkComponent
OptiMATE Dispense Probe
OptiMATE Dispense Probe
OptiMATE probe
Catalog # BECMD12222
New Product
Supplier:   Beckman Coulter
OptiMATE Dispense Probe
Catalog # BECMD12222
Supplier:  Beckman Coulter
New Product
OptiMATE Dispense Probe
OptiMATE Dispense Probe
OptiMATE probe
Price
120,00 €
Log in for availability
1 * 10 items
Product Details & Documents
The OptiMATE Dispense Probe is connected to the mixing manifold (a part of the upper fluidics) to enable the dispensing of reagents. Contains 10 units.
Specifications
  • Description:
    OptiMATE probe