Orders LinkContactUsLinkComponent
OptiMATE Probe Guide
OptiMATE Probe Guide
OptiMATE probe guide
Catalog # BECMD12218
New Product
Supplier:   Beckman Coulter
OptiMATE Probe Guide
Catalog # BECMD12218
Supplier:  Beckman Coulter
New Product
OptiMATE Probe Guide
OptiMATE Probe Guide
OptiMATE probe guide
Price
360,50 €
Log in for availability
1 * 4 items
Product Details & Documents
The OptiMATE Probe Guide aligns the probe and the tubes during the dispensing of reagents. Contains 4 units. Refer to OptiMATE Gradient Maker IFU for proper usage of the consumables.
Specifications
  • Description:
    OptiMATE probe guide