Orders LinkContactUsLinkComponent
OptiMATE Upper Fluidics
OptiMATE Upper Fluidics
OptiMATE upper fluidics
Catalog # BECMD12216
New Product
Supplier:   Beckman Coulter
OptiMATE Upper Fluidics
Catalog # BECMD12216
Supplier:  Beckman Coulter
New Product
OptiMATE Upper Fluidics
OptiMATE Upper Fluidics
OptiMATE upper fluidics
Price
409,60 €
Log in for availability
1 * 2 KIT
Product Details & Documents
The OptiMATE Upper Fluidics is the fluid path that enables reagent pumping via peristaltic pumps, mixing in the manifold, and dispensing via the probe into the tubes. Contains 2 units. Refer to OptiMATE Gradient Maker IFU for proper usage of the consumables. Endotoxin: ≤10 EU/device.
Specifications
  • Description:
    OptiMATE upper fluidics