Orders LinkContactUsLinkComponent
OptiMATE Flush Kit
OptiMATE  flush kit
  BECMD12215
New Product
OptiMATE Flush Kit
  BECMD12215
New Product
OptiMATE Flush Kit
OptiMATE Flush Kit
OptiMATE  flush kit
233,00 €
1 * 4 KIT
The OptiMATE Flush kit is needed for cleaning of the fluidic path, ie., OptiMATE Upper Fluidics. Proper cleaning will ensure the upper fluidics is ready to use the next time you need it.
  • Description:
    OptiMATE  flush kit