OptiMATE Iodixanol Solution
OptiMATE iodixanol reagent
Catalog # BECMD01358
New Product
Supplier: Beckman Coulter
Jump to:
Product Details & Documents
Iodixanol for your purification needs. Ideal for step gradients common to viral, organelle, EV, and protein separations. Concentrated to 80%, OptiMATE Iodixanol can enable you to increase your sample per tube and decrease your bottle-related waste.
Volume: 500 ml
Density: 1,420 ±0,004 g/ml
Concentration: 80% w/v
Refractive Index: 1,460 ±0,004
Endotoxin Level: ≤1 EU/ml
Sterile: Yes (autoclaved)
Shelf Life: 18 months
Bottle Dimensions (H×W): 17,6×7,2 cm (6,93×2,83")
Specifications
- Description:OptiMATE iodixanol reagent



