OptiMATE Cesium Chloride Solution
OptiMATE cesium chloride reagent
Catalog # BECMD01357
New Product
Supplier: Beckman Coulter
Jump to:
Product Details & Documents
Highly concentrated, premixed CsCl solution offering significant time savings in reagent preparation. Ideal for continuous gradients requiring the highest quality purification, commonly used in viral, plasmid, or nucleic acid purification.
Volume: 500 ml
Density: 1,784 ±0,014 g/ml
Concentration: 60% w/w
Refractive Index: 1,408 ±0,004
Endotoxin Level: ≤1 EU/ml
Sterile: Yes (autoclaved)
Shelf Life: 18 months
Bottle Dimensions (H×W): 17,6×7,2 cm (6,93×2,83")
Specifications
- Description:OptiMATE cesium chloride reagent



