0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Lecithin
BP0000735285
L-α-Phosphatidylcholine
New Product
:
Apollo Scientific
L-α-Phosphatidylcholine
:
Apollo Scientific
New Product
L-α-Phosphatidylcholine
Product Family Options
Pack type
APOSOR1028041-2.5K
Bottle
1 * 2,5 kg
1 * 2,5 kg - 173,00 €
173,00 €
-
+
1 * 2,5 kg
173,00 €
-
+
New Product
New Product
APOSOR1028041-100G
Bottle
1 * 100 g
1 * 100 g - 24,60 €
24,60 €
-
+
1 * 100 g
24,60 €
-
+
New Product
New Product
APOSOR1028041-25G
Bottle
1 * 25 g
1 * 25 g - 21,80 €
21,80 €
-
+
1 * 25 g
21,80 €
-
+
New Product
New Product
APOSOR1028041-500G
Bottle
1 * 500 g
1 * 500 g - 37,70 €
37,70 €
-
+
1 * 500 g
37,70 €
-
+
New Product
New Product