Order Entry
Export
Orders LinkContactUsLinkComponent
 

Tetracosane 99%

Supplier:   Apollo Scientific
Tetracosane 99%
Supplier:  Apollo Scientific
Certificates

5 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent

  • Non-hazardous
Synonyms:  N-tetracosane
  • CAS Number:646-31-1
  • Formula:C₂₄H₅₀
  • MDL Number:MFCD00009352
  • Molecular Weight:338,65 g/mol
  • EINECS:211-474-5
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 73,80 €
    73,80 €
    1 * 25 g
       73,80 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 228,00 €
    228,00 €
    1 * 100 g
       228,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 090,00 €
    1 090,00 €
    1 * 500 g
       1 090,00 €