Order Entry
Export
Orders LinkContactUsLinkComponent
N-Tetradecanamide 97%

N-Tetradecanamide 97%

New Product
Supplier:   Apollo Scientific
N-Tetradecanamide 97%
Supplier:  Apollo Scientific
New Product
Certificates
N-Tetradecanamide 97%

6 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  N-Tetradecanamide
  • CAS Number:638-58-4
  • Formula:C₁₄H₂₉NO
  • MDL Number:MFCD00025533
  • Molecular Weight:227,39 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 21,80 €
    21,80 €
    1 * 250 mg
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 24,60 €
    24,60 €
    1 * 1 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 30,40 €
    30,40 €
    1 * 5 g
       30,40 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 150,00 €
    150,00 €
    1 * 25 g
       150,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 582,00 €
    582,00 €
    1 * 100 g
       582,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 1 570,00 €
    1 570,00 €
    1 * 500 g
       1 570,00 €

    Specifications:
    New Product
    New Product