Order Entry
Export
Orders LinkContactUsLinkComponent
AC-Beta-ALA-OH 97%

AC-Beta-ALA-OH 97%

Supplier:   Apollo Scientific
AC-Beta-ALA-OH 97%
Supplier:  Apollo Scientific
Certificates
AC-Beta-ALA-OH 97%

6 Options Available Below

Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  N-Ac-β-Ala-OH, 3-Acetamidopropanoic acid, N-Acetyl-beta-alanine, N-Acetyl-b-alanine
  • CAS Number:3025-95-4
  • Formula:C₅H₉NO₃
  • MDL Number:MFCD00037296
  • Molecular Weight:131,13 g/mol
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 24,60 €
    24,60 €
    1 * 5 g
       24,60 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 30,40 €
    30,40 €
    1 * 25 g
       30,40 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 126,00 €
    126,00 €
    1 * 100 g
       126,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 394,00 €
    394,00 €
    1 * 500 g
       394,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 1 950,00 €
    1 950,00 €
    1 * 2,5 kg
       1 950,00 €

    Specifications:
    New Product
    New Product