Order Entry
Export
Orders LinkContactUsLinkComponent
Pyrazinamine ≥98%

Pyrazinamine ≥98%

Bottle
Catalog # APOSBUP19391-1G
New Product
Supplier:   Apollo Scientific
Pyrazinamine ≥98%
Catalog # APOSBUP19391-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  5049-61-6
Pyrazinamine ≥98%

Bottle

CAS Number:  5049-61-6
Price
37,70 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  Aminopyrazine, Pyrazin-2-amine
  • CAS Number:5049-61-6
  • Formula:C₄H₅N₃
  • Molecular Weight:95,1 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications