Order Entry
Export
Orders LinkContactUsLinkComponent
1-Naphthol ≥95%

1-Naphthol ≥95%

Bottle
Catalog # APOSBUP19222-1G
New Product
Supplier:   Apollo Scientific
1-Naphthol ≥95%
Catalog # APOSBUP19222-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  90-15-3
1-Naphthol ≥95%

Bottle

CAS Number:  90-15-3
Price
37,70 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/life science applications

Synonyms:  1-Hydroxynaphthalene, α-Naphthol, α-Hydroxynaphthalene
  • CAS Number:90-15-3
  • Formula:C₁₀H₈O
  • Molecular Weight:144,17 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications