Order Entry
Export
Orders LinkContactUsLinkComponent
4,4'-DIMETHOXYBENZIL ≥98%

4,4'-DIMETHOXYBENZIL ≥98%

Bottle
  APOSBUP19079-1G
New Product
4,4'-DIMETHOXYBENZIL ≥98%
  APOSBUP19079-1G
New Product
 :  1226-42-2
4,4'-DIMETHOXYBENZIL ≥98%

Bottle

 :  1226-42-2
 
37,70 €
Log in for availability
1 * 1 g
 

Usage: Biochemical/life science applications

Synonyms:  Bis(4-methoxyphenyl)ethanedione, Anisil, p-Anisil, 1,2-Bis(4-methoxyphenyl)ethane-1,2-dione
  • CAS Number:1226-42-2
  • Formula:C₁₆H₁₄O₄
  • Molecular Weight:270,28 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications