Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Taurine
BP0000733438
Taurine
Print
Taurine
New Product
Supplier:
Apollo Scientific
Taurine
Supplier:
Apollo Scientific
New Product
Certificates
Expand image
Return to Top
Taurine
Product Family Options
Add to cart
Jump to:
Product Family Options
About this item
2 Options Available Below
Shop Now
Return Policy
Product Details & Documents
Product Family Options
Product Information
Pack type
Availability
Price
APOSBUP18800-500MG
Bottle
Log in
for availability
Log in
for availability
1 * 500 mg
1 * 500 mg - 52,10 €
52,10 €
-
+
1 * 500 mg
52,10 €
-
+
Specifications:
New Product
New Product
APOSBUP18800-1G
Bottle
Log in
for availability
Log in
for availability
1 * 1 g
1 * 1 g - 62,30 €
62,30 €
-
+
1 * 1 g
62,30 €
-
+
Specifications:
New Product
New Product